Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dimethyl-1H-1,2,3-triazole 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A151431-100MG A151431-1G A151431-250MG A151431-5G
77010-806EA 13 USD InStock
77010-806 77010-808 77010-810 77683-277
1,4-Dimethyl-1H-1,2,3-triazole 95%, Ambeed.Inc
1,4-Dimethyl-1H-1,2,3-triazole
Precaución: For research use only.


Formula: C₄H₇N₃
MW: 97.11 g/mol
MDL Number: MFCD25976433
CAS Number: 60166-43-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty