Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Difluoro-2-nitrophenol 98%

Supplier: AMBEED, INC
Danger

A151548-100MG A151548-10G A151548-1G A151548-250MG A151548-5G
77010-828EA 56 USD InStock
77010-828 77010-830 77010-832 77010-834 77010-836
3,6-Difluoro-2-nitrophenol 98%
3,6-Difluoro-2-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₃F₂NO₃
MDL Number: MFCD22042170
CAS Number: 139548-97-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty