Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(S)-3-amino-4-(3,4-difluorophenyl)butyric acid 98%

Supplier: AMBEED, INC
Warning

A151810-1G A151810-250MG
77028-144EA 122 USD InStock
77028-144 77028-146
Fmoc-(S)-3-amino-4-(3,4-difluorophenyl)butyric acid 98%
Fmoc-(S)-3-amino-4-(3,4-difluorophenyl)butyric acid
Precaución: for Research Use Only


Formula: C₂₅H₂₁F₂NO₄
MDL Number: MFCD01861051
CAS Number: 270063-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty