Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-aminohexanoate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A152184-100MG A152184-250MG
76984-672EA 8 USD InStock
76984-672 76984-674
(S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-aminohexanoate hydrochloride 97%
(S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-aminohexanoate hydrochloride
Precaución: for Research Use Only


Formula: C₂₂H₂₆N₂O₄·ClH
MDL Number: MFCD08275774
CAS Number: 847658-45-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty