Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-hydroxybenzotrifluoride 98%

Supplier: AMBEED, INC
Warning

A152260-100MG A152260-10G A152260-1G A152260-250MG A152260-25G
77028-188EA 5 USD InStock
77028-188 77028-190 77028-192 77028-194 77028-196
2-Amino-5-hydroxybenzotrifluoride 98%
2-Amino-5-hydroxybenzotrifluoride
Precaución: for Research Use Only


Formula: C₇H₆F₃NO
MDL Number: MFCD01862068
CAS Number: 445-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty