Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(N-γ-4-methyltrityl)-L-α,γ-diaminobutyric acid 95%

Supplier: AMBEED, INC
Warning

A152378-1G A152378-250MG A152378-5G
77010-858EA 24 USD InStock
77010-858 77010-860 77010-862
Fmoc-(N-γ-4-methyltrityl)-L-α,γ-diaminobutyric acid 95%
Fmoc-(N-gamma-4-methyltrityl)-L-alpha,gamma-diaminobutyric acid
Precaución: for Research Use Only


Formula: C₃₉H₃₆N₂O₄
MW: 596.73 g/mol
MDL Number: MFCD02094096
CAS Number: 851392-68-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty