Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-methyl-6-nitrobenzoate 97%

Supplier: AMBEED, INC
Warning

A152423-100G A152423-10G A152423-1G A152423-25G A152423-5G
77028-200EA 945 USD InStock
77028-200 77028-202 77028-204 77028-206 77028-208
Methyl 2-methyl-6-nitrobenzoate 97%
Methyl 2-methyl-6-nitrobenzoate
Precaución: for Research Use Only


Formula: C₉H₉NO₄
MDL Number: MFCD11042453
CAS Number: 61940-22-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty