Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate 98%

Supplier: AMBEED, INC
Warning

A152465-25G A152465-5G A152465-100G
76997-392EA 7.43 USD InStock
76997-392 76997-394 76997-396 77549-920
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate 98%
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
Precaución: For research use only.


Formula: C₅H₁₂ClF₆N₂P
MDL Number: MFCD01862891
CAS Number: 94790-35-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty