Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-(4-fluorobenzyloxy)-5-methylphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A152997-100MG A152997-1G A152997-250MG
77010-896EA 11 USD InStock
77010-896 77010-898 77010-900
3-Bromo-2-(4-fluorobenzyloxy)-5-methylphenylboronic acid 97%
3-Bromo-2-(4-fluorobenzyloxy)-5-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₃BBrFO₃
MW: 338.9700 g/mol
MDL Number: MFCD06411360
CAS Number: 849062-41-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty