Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Piperidineethanol 97%

Supplier: AMBEED, INC
Danger

A153931-10G A153931-25G A153931-5G
76997-458EA 7 USD InStock
76997-458 76997-460 76997-462
2-Piperidineethanol 97%
2-Piperidineethanol
Precaución: for Research Use Only


Formula: C₇H₁₅NO
MDL Number: MFCD00005989
CAS Number: 1484-84-0
UN: 3263
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty