Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-Tyr(Et)-OH 98%

Supplier: AMBEED, INC
Warning

A154053-10G A154053-250MG
77028-284EA 84 USD InStock
77028-284 77028-286
Boc-D-Tyr(Et)-OH 98%
Boc-D-Tyr(Et)-OH
Precaución: for Research Use Only


Formula: C₁₆H₂₃NO₅
MDL Number: MFCD00065602
CAS Number: 76757-92-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty