Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl ((4-bromopyridin-2-yl)methyl)carbamate 97%

Supplier: AMBEED, INC
Warning

A154107-100MG A154107-250MG
77010-936EA 23 USD InStock
77010-936 77010-938
tert-Butyl ((4-bromopyridin-2-yl)methyl)carbamate 97%
tert-Butyl ((4-bromopyridin-2-yl)methyl)carbamate
Precaución: for Research Use Only


Formula: C₁₁H₁₅BrN₂O₂
MDL Number: MFCD13189516
CAS Number: 1060813-12-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty