Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-bromo-3-(trifluoromethyl)picolinate 98%

Supplier: AMBEED, INC
Warning

A154313-100MG A154313-1G A154313-250MG A154313-5G
77028-294EA 24 USD InStock
77028-294 77028-296 77028-298 77028-300
Methyl-5-bromo-3-(trifluoromethyl)picolinate 98%
Methyl-5-bromo-3-(trifluoromethyl)picolinate
Precaución: for Research Use Only


Formula: C₈H₅BrF₃NO₂
MW: 284.0300 g/mol
MDL Number: MFCD14698001
CAS Number: 1214328-84-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty