Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methylpyrazine-2-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A154840-100MG A154840-1G A154840-250MG A154840-5G
76997-476EA 19 USD InStock
76997-476 76997-478 76997-480 76997-482
6-Methylpyrazine-2-carbaldehyde 95%
6-Methylpyrazine-2-carbaldehyde
Precaución: for Research Use Only


Formula: C₆H₆N₂O
MDL Number: MFCD10700255
CAS Number: 116758-01-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty