Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo[1,2,4]triazolo[1,5-a]pyrazine 95%

Supplier: AMBEED, INC
Warning

A156136-100MG A156136-1G A156136-250MG A156136-5G
77028-370EA 26 USD InStock
77028-370 77028-372 77028-374 77028-376
6-Bromo[1,2,4]triazolo[1,5-a]pyrazine 95%
6-Bromo[1,2,4]triazolo[1,5-a]pyrazine
Precaución: for Research Use Only


Formula: C₅H₃BrN₄
MDL Number: MFCD12923113
CAS Number: 1233026-51-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty