Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ABT-199 99%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A156386-1MG A156386-250MG A156386-2MG A156386-25G
77010-994EA 14 USD InStock
77010-994 77010-996 77010-998 77683-307
ABT-199 99%, Ambeed.Inc
ABT-199
Precaución: For research use only.


Formula: C₄₅H₅₀ClN₇O₇S
MDL Number: MFCD23160052
CAS Number: 1257044-40-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty