Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitro-α,α,α-trifluoro-o-toluic acid 98%

Supplier: AMBEED, INC
Warning

A156788-10G A156788-25G A156788-1G
76997-564EA 28 USD InStock
76997-564 76997-566 77671-212
4-Nitro-α,α,α-trifluoro-o-toluic acid 98%
4-Nitro-α,α,α-trifluoro-o-toluic acid
Precaución: for Research Use Only


Formula: C₈H₄F₃NO₄
MW: 235.12
MDL Number: MFCD04039246
CAS Number: 320-37-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty