Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Furoyltrifluoroacetone 98%

Supplier: AMBEED, INC
Warning

A158401-100G A158401-10G A158401-1G A158401-25G A158401-5G
76984-944EA 65 USD InStock
76984-944 76984-946 76984-948 76984-950 76984-952
2-Furoyltrifluoroacetone 98%
2-Furoyltrifluoroacetone
Precaución: for Research Use Only


Formula: C₈H₅F₃O₃
MDL Number: MFCD00020935
CAS Number: 326-90-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty