Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-5-chlorophenylboronic acid 96%

Supplier: AMBEED, INC
Warning

A158527-10G A158527-250MG A158527-25G A158527-5G A158527-1G
76997-664EA 140 USD InStock
76997-664 76997-666 76997-668 76997-670 77671-244
2-Benzyloxy-5-chlorophenylboronic acid 96%
2-Benzyloxy-5-chlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₂BClO₃
MW: 262.4966 g/mol
MDL Number: MFCD04039000
CAS Number: 612832-83-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty