Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Dichloropyridazine 98%

Supplier: AMBEED, INC
Warning

A160468-100G A160468-500G A160468-25G
76997-778EA 13 USD InStock
76997-778 76997-780 76997-782 77300-470
3,6-Dichloropyridazine 98%
3,6-Dichloropyridazine
Precaución: for Research Use Only


Formula: C₄H₂Cl₂N₂
MDL Number: MFCD00006466
CAS Number: 141-30-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty