Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-pyridinesulfonamide 98%

Supplier: AMBEED, INC
Warning

A161235-100G A161235-10G A161235-1G A161235-250MG
77011-208EA 115 USD InStock
77011-208 77011-210 77011-212 77011-214
4-Chloro-3-pyridinesulfonamide 98%
4-Chloro-3-pyridinesulfonamide
Precaución: for Research Use Only


Formula: C₅H₅ClN₂O₂S
MW: 192.62 g/mol
MDL Number: MFCD04038136
CAS Number: 33263-43-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty