Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrakis(hydroxymethyl)phosphonium chloride in water Ca.80%

Supplier: AMBEED, INC
Danger

A161877-25G A161877-100G
76997-838EA 6 USD InStock
76997-838 77600-296
Tetrakis(hydroxymethyl)phosphonium chloride in water Ca.80%
Tetrakis(hydroxymethyl)phosphonium chloride
Precaución: for Research Use Only


Formula: C₄H₁₂ClO₄P
MDL Number: MFCD00031687
CAS Number: 124-64-1
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty