Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-nitrophenol 98%

Supplier: AMBEED, INC
Danger

A162176-100MG A162176-10G A162176-1G A162176-250MG A162176-25G A162176-5G
77011-248EA 6 USD InStock
77011-248 77011-250 77011-252 77011-254 77011-256 77011-258
2-Bromo-3-nitrophenol 98%
2-Bromo-3-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₄BrNO₃
MDL Number: MFCD08704555
CAS Number: 101935-40-4
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty