Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A162213-100MG A162213-10G A162213-1G A162213-250MG
77011-262EA 13 USD InStock
77011-262 77011-264 77011-266 77683-340
7-Bromo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one 97%, Ambeed.Inc
7-Bromo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
Precaución: For research use only.


Formula: C₇H₅BrN₂O₂
MDL Number: MFCD01648627
CAS Number: 105544-36-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty