Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-methyl-3-oxohexanoate 97% (by GC)

Supplier: AMBEED, INC
Warning

A162535-1G A162535-250MG A162535-25G A162535-5G
76985-100EA 21 USD InStock
76985-100 76985-102 76985-104 76985-106
Methyl 5-methyl-3-oxohexanoate 97% (by GC)
Methyl 5-methyl-3-oxohexanoate
Precaución: for Research Use Only


Formula: C₈H₁₄O₃
MDL Number: MFCD12031307
CAS Number: 30414-55-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty