Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

EPZ004777 98%

Supplier: AMBEED, INC
Warning

A162690-10MG A162690-1MG A162690-5MG
77011-284EA 385 USD InStock
77011-284 77011-286 77011-288
EPZ004777 98%
EPZ004777
Precaución: for Research Use Only


Formula: C₂₈H₄₁N₇O₄
MW: 539.67 g/mol
MDL Number: MFCD25563264
CAS Number: 1338466-77-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty