Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-amino-5-chloro-2-methoxybenzoate 96%

Supplier: AMBEED, INC
Warning

A163739-1G A163739-250MG A163739-5G
77011-326EA 70 USD InStock
77011-326 77011-328 77011-330
Methyl 4-amino-5-chloro-2-methoxybenzoate 96%
Methyl 4-amino-5-chloro-2-methoxybenzoate
Precaución: for Research Use Only


Formula: C₉H₁₀ClNO₃
MW: 215.634 g/mol
MDL Number: MFCD01034555
CAS Number: 20896-27-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty