Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Ethoxy-1H-indole-3-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A164326-100MG A164326-1G A164326-250MG A164326-5G
76997-936EA 13 USD InStock
76997-936 76997-938 76997-940 76997-942
5-Ethoxy-1H-indole-3-carbaldehyde 98%
5-Ethoxy-1H-indole-3-carbaldehyde
Precaución: for Research Use Only


Formula: C11H11NO2
MW: 189.2100 g/mol
MDL Number: MFCD07364715
CAS Number: 169789-47-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty