Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromothieno[2,3-b]pyridine 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A165936-250MG A165936-1G
76985-222EA 48 USD InStock
76985-222 77683-359
5-Bromothieno[2,3-b]pyridine 98%, Ambeed.Inc
5-Bromothieno[2,3-b]pyridine
Precaución: For research use only.


Formula: C₇H₄BrNS
MDL Number: MFCD18254753
CAS Number: 21344-24-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty