Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-chloro-2-methylbenzoate 97%

Supplier: AMBEED, INC
Warning

A166742-1G A166742-5G
77011-436EA 53 USD InStock
77011-436 77011-438
Ethyl 5-chloro-2-methylbenzoate 97%
Ethyl 5-chloro-2-methylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₁ClO₂
MDL Number: MFCD11226198
CAS Number: 56427-54-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty