Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4,6-difluorobenzothiazole 95%

Supplier: AMBEED, INC
Warning

A167614-10G A167614-1G A167614-250MG
76998-072EA 131 USD InStock
76998-072 76998-074 76998-076
2-Chloro-4,6-difluorobenzothiazole 95%
2-Chloro-4,6-difluorobenzothiazole
Precaución: for Research Use Only


Formula: C₇H₂ClF₂NS
MDL Number: MFCD04971835
CAS Number: 252681-57-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty