Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,3-dioxo-1,2,3,4-tetrahydro-6-quinoxalinecarboxylate 95%

Supplier: AMBEED, INC
Warning

A168594-100MG A168594-1G A168594-5G
76998-150EA 22 USD InStock
76998-150 76998-152 76998-154
Methyl-2,3-dioxo-1,2,3,4-tetrahydro-6-quinoxalinecarboxylate 95%
Methyl-2,3-dioxo-1,2,3,4-tetrahydro-6-quinoxalinecarboxylate
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O₄
MDL Number: MFCD11973607
CAS Number: 354793-04-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty