Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-indazole-5-carboxaldehyde 97%

Supplier: AMBEED, INC
Warning

A169496-100MG A169496-10G A169496-1G A169496-250MG A169496-5G
77028-956EA 20 USD InStock
77028-956 77028-958 77028-960 77028-962 77028-964
1-Methyl-1H-indazole-5-carboxaldehyde 97%
1-Methyl-1H-indazole-5-carboxaldehyde
Precaución: for Research Use Only


Formula: C₉H₈N₂O
MDL Number: MFCD11109365
CAS Number: 872607-89-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty