Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(2-Fluorophenyl)nicotinic acid 95%

Supplier: AMBEED, INC
Warning

A169587-100MG A169587-1G A169587-250MG
77028-966EA 79 USD InStock
77028-966 77028-968 77028-970
5-(2-Fluorophenyl)nicotinic acid 95%
5-(2-Fluorophenyl)nicotinic acid
Precaución: for Research Use Only


Formula: C₁₂H₈FNO₂
MW: 217.196 g/mol
MDL Number: MFCD06410258
CAS Number: 705961-96-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty