Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,3R)-3-(((Benzyloxy)carbonyl)amino)-2-hydroxy-4-phenylbutanoic acid 95%

Supplier: AMBEED, INC
Warning

A171625-250MG A171625-5G
76985-450EA 102.91 USD InStock
76985-450 76985-452 76985-454
(2S,3R)-3-(((Benzyloxy)carbonyl)amino)-2-hydroxy-4-phenylbutanoic acid 95%
(2S,3R)-3-(((Benzyloxy)carbonyl)amino)-2-hydroxy-4-phenylbutanoic acid
Precaución: for Research Use Only


Formula: C₁₈H₁₉NO₅
MDL Number: MFCD23701663
CAS Number: 59969-65-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty