Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropyl cinnamate 98%

Supplier: AMBEED, INC
Warning

A172298-100G A172298-25G A172298-500G
77029-104EA 54 USD InStock
77029-104 77029-106 77029-108
Isopropyl cinnamate 98%
Isopropyl cinnamate
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
MDL Number: MFCD00048292
CAS Number: 7780-06-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty