Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-iodobenzonitrile 97%

Supplier: AMBEED, INC
Warning

A173840-100MG A173840-1G A173840-250MG A173840-5G
76985-560EA 23 USD InStock
76985-560 76985-562 76985-564 76985-566
3-Bromo-5-iodobenzonitrile 97%
3-Bromo-5-iodobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃BrIN
MW: 307.9139 g/mol
MDL Number: MFCD00672949
CAS Number: 289039-21-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty