Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ACY-1215 98+%

Supplier: AMBEED, INC
Warning

A173859-10MG A173859-1MG
77011-696EA 36 USD InStock
77011-696 77011-698
ACY-1215 98+%
ACY-1215
Precaución: for Research Use Only


Formula: C₂₄H₂₇N₅O₃
MDL Number: MFCD22666356
CAS Number: 1316214-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty