Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Di-n-octyl-4,4'-bipyridinium dibromide 95%

Supplier: AMBEED, INC
Warning

A174216-1G A174216-250MG A174216-5G
76998-316EA 35 USD InStock
76998-316 76998-318 76998-320
1,1'-Di-n-octyl-4,4'-bipyridinium dibromide 95%
1,1'-Di-n-octyl-4,4'-bipyridinium dibromide
Precaución: for Research Use Only


Formula: C₂₆H₄₂Br₂N₂
MDL Number: MFCD00060051
CAS Number: 36437-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty