Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-3,4-dichloro-D-phenylalanine 98%

Supplier: AMBEED, INC
Warning

A174348-1G A174348-250MG A174348-5G A174348-25G
76998-330EA 45 USD InStock
76998-330 76998-332 76998-334 77671-470
Boc-3,4-dichloro-D-phenylalanine 98%
Boc-3,4-dichloro-D-phenylalanine
Precaución: for Research Use Only


Formula: C₁₄H₁₇Cl₂NO₄
MW: 334.193 g/mol
MDL Number: MFCD00151887
CAS Number: 114873-13-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty