Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxy-3,4-dihydroisoquinolin-1(2H)-one 98%

Supplier: AMBEED, INC
Warning

A176523-100MG A176523-1G A176523-250MG
77011-784EA 91 USD InStock
77011-784 77011-786 77011-788
6-Hydroxy-3,4-dihydroisoquinolin-1(2H)-one 98%
6-Hydroxy-3,4-dihydroisoquinolin-1(2H)-one
Precaución: for Research Use Only


Formula: C₉H₉NO₂
MDL Number: MFCD08059297
CAS Number: 22245-98-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty