Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,1'-Biphenyl]-3-ylhydrazine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A176727-100MG A176727-250MG
77011-802EA 44 USD InStock
77011-802 77011-804
[1,1'-Biphenyl]-3-ylhydrazine hydrochloride 95%
[1,1'-Biphenyl]-3-ylhydrazine hydrochloride
Precaución: for Research Use Only


Formula: C₁₂H₁₃ClN₂
MDL Number: MFCD06738781
CAS Number: 109221-88-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty