Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxy-5,6,7,8-tetrahydronaphthalene-2-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A176886-100MG A176886-1G A176886-250MG
76985-634EA 58 USD InStock
76985-634 76985-636 76985-638
5-Hydroxy-5,6,7,8-tetrahydronaphthalene-2-carbonitrile 95%
5-Hydroxy-5,6,7,8-tetrahydronaphthalene-2-carbonitrile
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO
MDL Number: MFCD22571375
CAS Number: 1315479-99-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty