Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anthranilonitrile 98%

Supplier: AMBEED, INC
Danger

A177188-100G A177188-10G A177188-1G A177188-25G A177188-500G A177188-5G
77029-290EA 52 USD InStock
77029-290 77029-292 77029-294 77029-296 77029-298 77029-300
Anthranilonitrile 98%
Anthranilonitrile
Precaución: for Research Use Only


Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 268
Melting Pt: 47
MDL Number: MFCD00007631
CAS Number: 1885-29-6
UN: 3439
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty