Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-2,5-dicarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A177818-25G A177818-5G
77029-312EA 22 USD InStock
77029-312 77029-314
Pyridine-2,5-dicarboxylic acid 98%
Pyridine-2,5-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357
MDL Number: MFCD00006297
CAS Number: 100-26-5
Merck Index: 13,05177
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty