Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2,6-dichlorobenzene 95%

Supplier: AMBEED, INC
Warning

A179120-100G A179120-10G A179120-25G A179120-500G A179120-5G
76998-552EA 84 USD InStock
76998-552 76998-554 76998-556 76998-558 76998-560
1-Bromo-2,6-dichlorobenzene 95%
1-Bromo-2,6-dichlorobenzene
Precaución: for Research Use Only


Formula: C₆H₃BrCl₂
MDL Number: MFCD00000574
CAS Number: 19393-92-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty