Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromophenyl)acetic acid ethyl ester 98%

Supplier: AMBEED, INC
Warning

A179230-100G A179230-10G A179230-500G A179230-5G A179230-25G
76998-588EA 45 USD InStock
76998-588 76998-590 76998-592 76998-594 77083-214
(4-Bromophenyl)acetic acid ethyl ester 98%
(4-Bromophenyl)acetic acid ethyl ester
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₂
MW: 243.1
MDL Number: MFCD00016333
CAS Number: 14062-25-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty