Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitro-1-phenyl-1H-pyrazole-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A179317-100MG A179317-1G A179317-250MG
77011-922EA 18 USD InStock
77011-922 77011-924 77011-926
4-Nitro-1-phenyl-1H-pyrazole-3-carboxylic acid 98%
4-Nitro-1-phenyl-1H-pyrazole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₇N₃O₄
MDL Number: MFCD20484387
CAS Number: 701917-03-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty