Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methyl-1,2,3,4-tetrahydropyrido[2,3-b]pyrazine 95%

Supplier: AMBEED, INC
Warning

A179528-1G A179528-250MG
77011-942EA 110 USD InStock
77011-942 77011-944
6-Methyl-1,2,3,4-tetrahydropyrido[2,3-b]pyrazine 95%
6-Methyl-1,2,3,4-tetrahydropyrido[2,3-b]pyrazine
Precaución: for Research Use Only


Formula: C₈H₁₁N₃
MDL Number: MFCD13193481
CAS Number: 1260769-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty