Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-2-methylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A179828-10G A179828-1G A179828-5G
76985-724EA 117 USD InStock
76985-724 76985-726 76985-728
4-Cyano-2-methylbenzoic acid 98%
4-Cyano-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₇NO₂
MDL Number: MFCD14582960
CAS Number: 1975-53-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty